3-Chloro-6-piperidin-1-ylpyridazine 98%

Lieferant: Apollo Scientific

Synonyme: 3-Chloro-6-(piperidin-1-yl)pyridazine

OR959487-10G OR959487-1G OR959487-25G
APOSOR959487-10GEA 342 EUR
APOSOR959487-10G APOSOR959487-1G APOSOR959487-25G
3-Chloro-6-piperidin-1-ylpyridazine 98%
3-Chloro-6-piperidin-1-ylpyridazine
Formel: C₉H₁₂ClN₃
Molecular Weight: 197,67 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00574572
CAS-Nummer: 1722-11-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR