N-(4-Chloropyridin-2-yl)acetamide 98%
Lieferant: Apollo Scientific
Synonyme:
2-Acetamido-4-chloropyridine
OR959541-10G
OR959541-1G
OR959541-5G
APOSOR959541-10GEA
516
EUR
APOSOR959541-10G
APOSOR959541-1G
APOSOR959541-5G
N-(4-Chloropyridin-2-yl)acetamide 98%
N-(4-Chloropyridin-2-yl)acetamide
Formel:
C₇H₇ClN₂O Molecular Weight: 170,6 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03394624 CAS-Nummer: 245056-66-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...