(3-Bromo-4-pyridyl)methanol 98%
Lieferant: Apollo Scientific
Synonyme:
3-Bromo-4-pyridinemethanol, 3-Bromo-4-(hydroxymethyl)pyridine
OR959577-5G
APOSOR959577-5GEA
108
EUR
APOSOR959577-5G
(3-Bromo-4-pyridyl)methanol 98%
(3-Bromo-4-pyridyl)methanol
Formel:
C₆H₆BrNO Molecular Weight: 188,03 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11847285 CAS-Nummer: 146679-66-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...