(4-Brom-3-pyridinyl)methanol 98%

Lieferant: Apollo Scientific
Achtung

OR959659-1G OR959659-250MG
APOSOR959659-1GEA 522 EUR
APOSOR959659-1G APOSOR959659-250MG
(4-Brom-3-pyridinyl)methanol 98%
(4-Brom-3-pyridinyl)methanol
Formel: C₆H₆BrNO
Lagertemperatur: Kühlschrank
CAS-Nummer: 197007-87-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR