5-Brom-2-hydroxybenzonitril 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR9598-25G
OR9598-1G
OR9598-5G
APOSOR9598-25GEA
91.5
EUR
APOSOR9598-25G
APOSOR9598-1G
APOSOR9598-5G
5-Brom-2-hydroxybenzonitril 98%
5-Brom-2-hydroxybenzonitril
Formel:
C₇H₄BrNO Schmelzpunkt: 158…163 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143428 CAS-Nummer: 40530-18-5 EINECS: 254-958-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...