Quinolin-8-ylmethanol 98%

Lieferant: Apollo Scientific

OR959827-1G OR959827-5G
APOSOR959827-1GEA 270 EUR
APOSOR959827-1G APOSOR959827-5G
Quinolin-8-ylmethanol 98%
Quinolin-8-ylmethanol
Formel: C₁₀H₉NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00769488
CAS-Nummer: 16032-35-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR