1-Ethyl-3-iod-1H-pyrazol
Lieferant: Apollo Scientific
|
Achtung
|
OR959875-1G
OR959875-5G
APOSOR959875-1GEA
46.4
EUR
APOSOR959875-1G
APOSOR959875-5G
1-Ethyl-3-iod-1H-pyrazol
1-Ethyl-3-iodo-1H-pyrazole
Formel:
C₅H₇IN₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD12756564 CAS-Nummer: 1202781-34-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...