2-Isopropyl-2H-pyrazole-3-carbaldehyde 95%

Lieferant: Apollo Scientific

Synonyme: 1-Isopropyl-1H-pyrazole-5-carbaldehyde

OR959891-1G OR959891-5G
APOSOR959891-1GEA 84.1 EUR
APOSOR959891-1G APOSOR959891-5G
2-Isopropyl-2H-pyrazole-3-carbaldehyde 95%
2-Isopropyl-2H-pyrazole-3-carbaldehyde
Formel: C₇H₁₀N₂O
Molecular Weight: 138.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03130042
CAS-Nummer: 100305-93-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR