N-(2-Thienylmethyl)butan-1-amine 98%
Lieferant: Apollo Scientific
Synonyme:
Butyl(thiophen-2-ylmethyl)amine
OR959974-10G
OR959974-1G
OR959974-5G
APOSOR959974-10GEA
502
EUR
APOSOR959974-10G
APOSOR959974-1G
APOSOR959974-5G
N-(2-Thienylmethyl)butan-1-amine 98%
N-(2-Thienylmethyl)butan-1-amine
Formel:
C₉H₁₅NS Molecular Weight: 169,29 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05668044 CAS-Nummer: 58924-53-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...