2'-Brom-5'-methoxyacetophenon
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(2-Bromo-5-methoxyphenyl)ethanone
OR9600-1G
OR9600-5G
APOSOR9600-1GEA
68.3
EUR
APOSOR9600-1G
APOSOR9600-5G
2'-Brom-5'-methoxyacetophenon
2'-Brom-5'-methoxyacetophenon
Formel:
C₉H₉BrO₂ Molecular Weight: 229.0706 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02683853 CAS-Nummer: 6342-63-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...