[4-(Ethylsulphonyl)phenyl]amine 96%

Lieferant: Apollo Scientific

Synonyme: 4-(Ethanesulphonyl)aniline

OR960089-10G OR960089-1G OR960089-5G
APOSOR960089-10GEA 864 EUR
APOSOR960089-10G APOSOR960089-1G APOSOR960089-5G
[4-(Ethylsulphonyl)phenyl]amine 96%
[4-(Ethylsulphonyl)phenyl]amine
Formel: C₈H₁₁NO₂S
Molecular Weight: 185,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01670261
CAS-Nummer: 6334-01-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR