4,5,6,7-Tetrahydro-1-benzothiophene-3-carboxylic acid 97%
Lieferant: Apollo Scientific
Synonyme:
4,5,6,7-Tetrahydrobenzo[b]thiophene-3-carboxylic acid
OR960181-5G
APOSOR960181-5GEA
208
EUR
APOSOR960181-5G
4,5,6,7-Tetrahydro-1-benzothiophene-3-carboxylic acid 97%
4,5,6,7-Tetrahydro-1-benzothiophene-3-carboxylic acid
Formel:
C₉H₁₀O₂S Molecular Weight: 182,24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00652575 CAS-Nummer: 19156-54-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...