2-(N-Anilino)pyrimidine 97%

Lieferant: Apollo Scientific

Synonyme: N-Phenylpyrimidin-2-amine

OR960387-1G OR960387-5G
APOSOR960387-1GEA 98.6 EUR
APOSOR960387-1G APOSOR960387-5G
2-(N-Anilino)pyrimidine 97%
2-(N-Anilino)pyrimidine
Formel: C₁₀H₉N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01646270
CAS-Nummer: 57356-49-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR