3-Phenylcyclohexanon 96%
Lieferant: Apollo Scientific
OR960434-1G
OR960434-250MG
APOSOR960434-1GEA
359
EUR
APOSOR960434-1G
APOSOR960434-250MG
3-Phenylcyclohexanon 96%
3-Phenylcyclohexanone
Formel:
C₁₂H₁₄O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00209998 CAS-Nummer: 20795-53-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...