2-Bromocyclohex-2-en-1-one 97%

Lieferant: Apollo Scientific

OR960465-1G OR960465-5G
APOSOR960465-1GEA 261 EUR
APOSOR960465-1G APOSOR960465-5G
2-Bromocyclohex-2-en-1-one 97%
2-Bromocyclohex-2-en-1-one
Formel: C₆H₇BrO
Lagertemperatur: Raumtemperatur
MDL: MFCD11109363
CAS-Nummer: 50870-61-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR