N-Benzyloxycarbonyl-L-norleucin 97%
Lieferant: Apollo Scientific
Synonyme:
N-Cbz-L-norleucine, Z-Nle-OH
OR960529-1G
OR960529-25G
OR960529-5G
APOSOR960529-1GEA
95.7
EUR
APOSOR960529-1G
APOSOR960529-25G
APOSOR960529-5G
N-Benzyloxycarbonyl-L-norleucin 97%
N-Benzyloxycarbonyl-L-norleucin
Formel:
C₁₄H₁₉NO₄ Molecular Weight: 265,309 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063170 CAS-Nummer: 39608-30-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...