Fmoc-D-tyrosine 97%

Lieferant: Apollo Scientific

Synonyme: Fmoc-D-Tyr-OH, N-Fmoc-D-tyrosine

OR960611-25G
APOSOR960611-25GEA 237 EUR
APOSOR960611-25G
Fmoc-D-tyrosine 97%
Fmoc-D-tyrosine
Formel: C₂₄H₂₁NO₅
Molecular Weight: 403,434 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00171384
CAS-Nummer: 112883-29-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR