4-Chloro-6-methylthiopyrimidine 98%

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-6-(methylthio)pyrimidine

OR960614-5G
APOSOR960614-5GEA 111 EUR
APOSOR960614-5G
4-Chloro-6-methylthiopyrimidine 98%
4-Chloro-6-methylthiopyrimidine
Formel: C₅H₅ClN₂S
Molecular Weight: 160,63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09870036
CAS-Nummer: 89283-48-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR