4-Chlor-2-(tributylstannyl)pyridin 97%
Lieferant: Apollo Scientific
|
Gefahr
|
OR960624-1G
OR960624-250MG
APOSOR960624-1GEA
397
EUR
APOSOR960624-1G
APOSOR960624-250MG
4-Chlor-2-(tributylstannyl)pyridin 97%
4-Chlor-2-(tributylstannyl)pyridin
Formel:
C₁₇H₃₀ClNSn Lagertemperatur: Kühlschrank |
MDL:
MFCD09870067 CAS-Nummer: 1204580-71-1 UN: 2788 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...