Fmoc-Dap-OH 97%

Lieferant: Apollo Scientific

Synonyme: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-aminopropanoic acid

OR960676-5G
APOSOR960676-5GEA 119 EUR
APOSOR960676-5G
Fmoc-Dap-OH 97%
Fmoc-Dap-OH
Formel: C₁₈H₁₈N₂O₄
Molecular Weight: 326,352 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00237019
CAS-Nummer: 181954-34-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR