6-Chlor-3-methylimidazo[1,2-b]pyridazin 97%
Lieferant: Apollo Scientific
OR960690-1G
OR960690-250MG
OR960690-5G
APOSOR960690-1GEA
197
EUR
APOSOR960690-1G
APOSOR960690-250MG
APOSOR960690-5G
6-Chlor-3-methylimidazo[1,2-b]pyridazin 97%
6-Chlor-3-methylimidazo[1,2-b]pyridazin
Formel:
C₇H₆ClN₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09991804 CAS-Nummer: 137384-48-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...