Fmoc-N-Me-Phe-OH 97%
Lieferant: Apollo Scientific
Synonyme: N-Fmoc-N-methyl-L-phenylalanine, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-3-phenylpropanoic acid, Fmoc-N-Me-Phe-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-phenylalanine
OR960697-1G OR960697-25G OR960697-5G
APOSOR960697-1GEA 50.9 EUR
APOSOR960697-1G APOSOR960697-25G APOSOR960697-5G
Fmoc-N-Me-Phe-OH 97%
Fmoc-N-Me-Phe-OH
Formel: C₂₅H₂₃NO₄
Molecular Weight: 401.462 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00151938
CAS-Nummer: 77128-73-5