5,6-Dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile 95%

Lieferant: Apollo Scientific

OR960712-1G OR960712-250MG
APOSOR960712-1GEA 139 EUR
APOSOR960712-1G APOSOR960712-250MG
5,6-Dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile 95%
5,6-Dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile
Formel: C₈H₈N₂O
Molecular Weight: 148,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03004855
CAS-Nummer: 72716-80-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR