2-Amino-3-(4-methoxyphenyl)propanoic acid 97%
Lieferant: Apollo Scientific
Synonyme:
H-DL-Tyr(Me)-OH
OR960745-1G
OR960745-25G
OR960745-5G
APOSOR960745-1GEA
116
EUR
APOSOR960745-1G
APOSOR960745-25G
APOSOR960745-5G
2-Amino-3-(4-methoxyphenyl)propanoic acid 97%
2-Amino-3-(4-methoxyphenyl)propanoic acid
Formel:
C₁₀H₁₃NO₃ Molecular Weight: 195,218 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063071 CAS-Nummer: 7635-29-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...