Boc-D-Chg-OH 97%

Lieferant: Apollo Scientific

Synonyme: Boc-D-Chg-OH, N-(tert-Butoxycarbonyl)-D-2-cyclohexylglycine, N-Boc-2-cyclohexyl-D-glycine, Boc-cyclohexyl-D-Gly-OH, (R)-2-((tert-Butoxycarbonyl)amino)-2-cyclohexylacetic acid, BOC-D-α-cyclohexylglycine, N-Boc-D-2-cyclohexylglycine

OR960787-1G OR960787-25G OR960787-5G
APOSOR960787-1GEA 52.2 EUR
APOSOR960787-1G APOSOR960787-25G APOSOR960787-5G
Boc-D-Chg-OH 97%
Boc-D-Chg-OH
Formel: C₁₃H₂₃NO₄
Molecular Weight: 257.33 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00133629
CAS-Nummer: 70491-05-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR