Fmoc-N-Me-D-Ala-OH 97%

Lieferant: Apollo Scientific

Synonyme: Fmoc-N-methyl-D-alanine

OR960869-25G OR960869-5G
APOSOR960869-25GEA 81.2 EUR
APOSOR960869-25G APOSOR960869-5G
Fmoc-N-Me-D-Ala-OH 97%
Fmoc-N-Me-D-Ala-OH
Formel: C₁₉H₁₉NO₄
Molecular Weight: 325.364 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00153385
CAS-Nummer: 138774-92-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR