4-(Hydroxymethyl)-2-isopropylthiazole 95%
Lieferant: Apollo Scientific
Synonyme:
(2-Isopropylthiazol-4-yl)methanol
OR960873-1G
APOSOR960873-1GEA
120
EUR
APOSOR960873-1G
4-(Hydroxymethyl)-2-isopropylthiazole 95%
4-(Hydroxymethyl)-2-isopropylthiazole
Formel:
C₇H₁₁NOS Molecular Weight: 157,24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09870064 CAS-Nummer: 133047-45-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...