Fmoc-L-norvaline 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-norvaline, Fmoc-Nva-OH, N-Fmoc-L-norvaline

OR960882-100G
APOSOR960882-100GEA 101 EUR
APOSOR960882-100G
Fmoc-L-norvaline 97%
Fmoc-L-norvaline
Formel: C₂₀H₂₁NO₄
Molecular Weight: 339,391 g/mol
Schmelzpunkt: 151…155 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00155631
CAS-Nummer: 135112-28-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR