Isochroman-4-one 95%

Lieferant: Apollo Scientific

OR960895-1G OR960895-250MG
APOSOR960895-1GEA 184 EUR
APOSOR960895-1G APOSOR960895-250MG
Isochroman-4-one 95%
Isochroman-4-one
Formel: C₉H₈O₂
Molecular Weight: 148.1586 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09744058
CAS-Nummer: 20924-56-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR