Imidazo[1,2-b]pyridazin-6-ol 95%

Lieferant: Apollo Scientific

Synonyme: 6-Hydroxyimidazo[1,2-b]pyridazine

OR960896-1G OR960896-5G
APOSOR960896-1GEA 42.2 EUR
APOSOR960896-1G APOSOR960896-5G
Imidazo[1,2-b]pyridazin-6-ol 95%
Imidazo[1,2-b]pyridazin-6-ol
Formel: C₆H₅N₃O
Molecular Weight: 135,1200 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11040161
CAS-Nummer: 57470-54-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR