Methyl-7-azaindole-2-carboxylate 98%
Lieferant: Apollo Scientific
Synonyme:
Methyl 1H-pyrrolo[2,3-b]pyridine-2-carboxylate
OR960932-10G
OR960932-1G
OR960932-5G
APOSOR960932-10GEA
230
EUR
APOSOR960932-10G
APOSOR960932-1G
APOSOR960932-5G
Methyl-7-azaindole-2-carboxylate 98%
Methyl-7-azaindole-2-carboxylate
Formel:
C₉H₈N₂O₂ Molecular Weight: 176.18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08752938 CAS-Nummer: 394223-02-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...