Boc-Cha-OH 97%
Lieferant: Apollo Scientific
Synonyme:
(S)-2-((tert-Butoxycarbonyl)amino)-3-cyclohexylpropanoic acid, Boc-β-cyclohexyl-Ala-OH
OR960946-25G
APOSOR960946-25GEA
127
EUR
APOSOR960946-25G
Boc-Cha-OH 97%
Boc-Cha-OH
Formel:
C₁₄H₂₅NO₄ Molecular Weight: 271,357 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00076896 CAS-Nummer: 37736-82-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...