2-(Methylthio)pyrimidin-4-ol 98%

Lieferant: Apollo Scientific

OR960960-500G
APOSOR960960-500GEA 1200 EUR
APOSOR960960-500G
2-(Methylthio)pyrimidin-4-ol 98%
2-(Methylthio)pyrimidin-4-ol
Formel: C₅H₆N₂OS
Lagertemperatur: Raumtemperatur
CAS-Nummer: 124700-70-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR