(R)-2-Amino-3-(4-benzoylphenyl)propanoic acid 97%
Lieferant: Apollo Scientific
Synonyme:
H-p-Bz-D-Phe-OH, H-D-Bpa-OH
OR960998-1G
OR960998-5G
APOSOR960998-1GEA
214
EUR
APOSOR960998-1G
APOSOR960998-5G
(R)-2-Amino-3-(4-benzoylphenyl)propanoic acid 97%
(R)-2-Amino-3-(4-benzoylphenyl)propanoic acid
Formel:
C₁₆H₁₅NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00238101 CAS-Nummer: 201466-03-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...