H-Pro-NHEt·HCl 98%

Lieferant: Apollo Scientific

Synonyme: H-Pro-NHEt·HCl, (S)-N-Ethylpyrrolidine-2-carboxamide hydrochloride

OR961001-10G OR961001-1G OR961001-5G
APOSOR961001-10GEA 79.9 EUR
APOSOR961001-10G APOSOR961001-1G APOSOR961001-5G
H-Pro-NHEt·HCl 98%
H-Pro-NHEt·HCl
Formel: C₇H₁₅ClN₂O
Molecular Weight: 178,66 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00238159
CAS-Nummer: 58107-62-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR