N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-piperidone 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-Fmoc-4-piperidone
OR961088-25G
OR961088-5G
APOSOR961088-25GEA
279
EUR
APOSOR961088-25G
APOSOR961088-5G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-piperidone 98%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-piperidone
Formel:
C₂₀H₁₉NO₃ Molecular Weight: 321,38 g/mol Schmelzpunkt: 133 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00673794 CAS-Nummer: 204376-55-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...