Phenylacetylglutamine 96%

Lieferant: Apollo Scientific

Synonyme: (S)-5-Amino-5-oxo-2-(2-phenylacetamido)pentanoic acid, Phenylac-Gln-OH

OR961244-10G OR961244-5G
APOSOR961244-10GEA 102 EUR
APOSOR961244-10G APOSOR961244-5G
Phenylacetylglutamine 96%
Phenylacetylglutamine
Formel: C₁₃H₁₆N₂O₄
Molecular Weight: 264,281 g/mol
Schmelzpunkt: 85…87 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01725188
CAS-Nummer: 28047-15-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR