L-Glutamic acid g-methyl ester a-amide hydrochloride 98%
Lieferant: Apollo Scientific
Achtung
Synonyme: H-Glu(OMe)-NH₂·HCl
OR961350-1G OR961350-25G OR961350-5G
APOSOR961350-1GEA 75.4 EUR
APOSOR961350-1G APOSOR961350-25G APOSOR961350-5G
L-Glutamic acid g-methyl ester a-amide hydrochloride 98%
L-Glutamic acid g-methyl ester a-amide hydrochloride
Formel: C₆H₁₃ClN₂O₃
Molecular Weight: 196,631 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02094379
CAS-Nummer: 70830-50-1