Fmoc-Cys-OH 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-mercaptopropanoic acid, Fmoc-L-cysteine

OR961460-1G OR961460-5G OR961460-10G
APOSOR961460-1GEA 97.3 EUR
APOSOR961460-1G APOSOR961460-5G APOSOR961460-10G
Fmoc-Cys-OH 96%
Fmoc-Cys-OH
Formel: C₁₈H₁₇NO₄S
Molecular Weight: 343.4 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03788178
CAS-Nummer: 135248-89-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR