(R)-3-Amino-3-(4-chlorophenyl)propanoic acid 97%

Lieferant: Apollo Scientific

OR961526-1G OR961526-5G
APOSOR961526-1GEA 48 EUR
APOSOR961526-1G APOSOR961526-5G
(R)-3-Amino-3-(4-chlorophenyl)propanoic acid 97%
(R)-3-Amino-3-(4-chlorophenyl)propanoic acid
Formel: C₉H₁₀ClNO₂
Molecular Weight: 199,63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03840387
CAS-Nummer: 131690-61-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR