2-Chloro-6-methoxypyrazine 98%
Lieferant: Apollo Scientific
OR961527-10G
OR961527-1G
OR961527-25G
APOSOR961527-10GEA
108
EUR
APOSOR961527-10G
APOSOR961527-1G
APOSOR961527-25G
2-Chloro-6-methoxypyrazine 98%
2-Chloro-6-methoxypyrazine
Formel:
C₅H₅ClN₂O Molecular Weight: 144,5600 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06410937 CAS-Nummer: 33332-30-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...