Methyl-2-methylpropionimidate hydrochloride 97%

Lieferant: Apollo Scientific

Synonyme: Methyl isobutyrimidate hydrochloride

OR961545-1G OR961545-5G
APOSOR961545-1GEA 89.9 EUR
APOSOR961545-1G APOSOR961545-5G
Methyl-2-methylpropionimidate hydrochloride 97%
Methyl-2-methylpropionimidate hydrochloride
Formel: C₅H₁₂ClNO
Molecular Weight: 137.607 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03844704
CAS-Nummer: 39739-60-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR