(R)-3-Amino-3-(2-bromophenyl)propionic acid 97%
Lieferant: Apollo Scientific
OR961588-1G
OR961588-500MG
APOSOR961588-1GEA
348
EUR
APOSOR961588-1G
APOSOR961588-500MG
(R)-3-Amino-3-(2-bromophenyl)propionic acid 97%
(R)-3-Amino-3-(2-bromophenyl)propionic acid
Formel:
C₉H₁₀BrNO₂ Molecular Weight: 244,09 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04113641 CAS-Nummer: 737751-95-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...