H-D-Phe(2,4-Dime)-OH 97%
Lieferant: Apollo Scientific
Synonyme:
2,4-Dimethyl-D-phenylalanine
OR961781-1G
OR961781-500MG
APOSOR961781-1GEA
433
EUR
APOSOR961781-1G
APOSOR961781-500MG
H-D-Phe(2,4-Dime)-OH 97%
H-D-Phe(2,4-Dime)-OH
Formel:
C₁₁H₁₅NO₂ Molecular Weight: 193,2 g/mol Lagertemperatur: Raumtemperatur |
CAS-Nummer:
465500-97-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...