H-D-Phe(3-CN)-OH 97%

Lieferant: Apollo Scientific

Synonyme: 3-Cyano-D-phenylalanine

OR961957-5G
APOSOR961957-5GEA 109 EUR
APOSOR961957-5G
H-D-Phe(3-CN)-OH 97%
H-D-Phe(3-CN)-OH
Formel: C₁₀H₁₀N₂O₂
Molecular Weight: 190,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01631985
CAS-Nummer: 263396-43-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR