(R)-2-Amino-3-(3-bromophenyl)propanoic acid 97%
Lieferant: Apollo Scientific
Synonyme:
H-D-Phe(3-Br)-OH
OR962027-1G
OR962027-25G
OR962027-5G
APOSOR962027-1GEA
109
EUR
APOSOR962027-1G
APOSOR962027-25G
APOSOR962027-5G
(R)-2-Amino-3-(3-bromophenyl)propanoic acid 97%
(R)-2-Amino-3-(3-bromophenyl)propanoic acid
Formel:
C₉H₁₀BrNO₂ Molecular Weight: 244,1 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659100 CAS-Nummer: 99295-78-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...