6-Chloro-1H-benzoimidazole-2-carboxylic acid 95%
Lieferant: Apollo Scientific
OR962177-1G
OR962177-5G
APOSOR962177-1GEA
255
EUR
APOSOR962177-1G
APOSOR962177-5G
6-Chloro-1H-benzoimidazole-2-carboxylic acid 95%
6-Chloro-1H-benzoimidazole-2-carboxylic acid
Formel:
C₈H₅ClN₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06739053 CAS-Nummer: 39811-14-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...