Fmoc-4-amino-tetrahydropyran-4-carboxylic acid 97%

Lieferant: Apollo Scientific

Synonyme: 4-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)tetrahydro-2H-pyran-4-carboxylic acid

OR962197-1G
APOSOR962197-1GEA 27.7 EUR
APOSOR962197-1G
Fmoc-4-amino-tetrahydropyran-4-carboxylic acid 97%
Fmoc-4-amino-tetrahydropyran-4-carboxylic acid
Formel: C₂₁H₂₁NO₅
Molecular Weight: 367,41 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02683137
CAS-Nummer: 285996-72-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR