1H-Indazol-1-ylacetic acid 95%

Lieferant: Apollo Scientific

OR962216-1G OR962216-5G
APOSOR962216-1GEA 197 EUR
APOSOR962216-1G APOSOR962216-5G
1H-Indazol-1-ylacetic acid 95%
1H-Indazol-1-ylacetic acid
Formel: C₉H₈N₂O₂
Molecular Weight: 176,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02938037
CAS-Nummer: 32829-25-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR