(S)-N-Fmoc-azetidine-2-carboxylic acid 97%

Lieferant: Apollo Scientific

Synonyme: (S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)azetidine-2-carboxylic acid, Fmoc-L-azetidine-2-carboxylic acid

OR962312-1G
APOSOR962312-1GEA 55.1 EUR
APOSOR962312-1G
(S)-N-Fmoc-azetidine-2-carboxylic acid 97%
(S)-N-Fmoc-azetidine-2-carboxylic acid
Formel: C₁₉H₁₇NO₄
Molecular Weight: 323,348 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01320843
CAS-Nummer: 136552-06-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR